Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023755.2 | NP_076244.2 | 5 - 6 | 598 | 111 | |
GenBank mRNA | AF311309.1 | - | 5 - 6 | 593 | 111 | |
AK143960.1 | - | 5 - 6 | 597 | 111 | ||
BC116663.1 | - | 5 - 6 | 593 | 40087602 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |