Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slamf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030710.2 | NP_109635.1 | 5 - 6 | 1004 | 90 | |
GenBank mRNA | AB196808.1 | - | 5 - 6 | 833 | 90 | |
AB196809.1 | - | 5 - 6 | 833 | 90 | ||
AB196810.1 | - | 5 - 6 | 833 | 90 | ||
AB196811.1 | - | 5 - 6 | 833 | 90 | ||
AB196836.1 | - | 5 - 6 | 833 | 90 | ||
AF248635.1 | - | 5 - 6 | 1004 | 90 | ||
AF248636.1 | - | 5 - 6 | 1026 | 90 | ||
BC030031.1 | - | 5 - 6 | 985 | 1225313 | 90 | |
EU591721.1 | - | 5 - 6 | 833 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |