Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021607.3 | NP_067620.3 | 1 - 2 | 154 | 127 | |
GenBank mRNA | AF240469.1 | - | 1 - 2 | 104 | 127 | |
AK033314.1 | - | 1 - 2 | 96 | 127 | ||
AK129102.1 | - | 1 - 2 | 154 | 127 | ||
AK170173.1 | - | 1 - 2 | 100 | 128 | ||
AK170244.1 | - | 1 - 2 | 119 | 127 | ||
AK170369.1 | - | 1 - 2 | 143 | 127 | ||
BC019998.1 | - | 1 - 2 | 99 | 4458780 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |