Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009813.2 | NP_033943.2 | 5 - 6 | 856 | 88 | |
GenBank mRNA | BC064057.1 | - | 5 - 6 | 794 | 876739 | 88 |
CR756967.1 | - | 2 - 3 | 88 | |||
U93291.1 | - | 5 - 6 | 812 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |