Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160049.1 | NP_001153521.1 | 1 - 2 | 106 | 101 | |
GenBank mRNA | AK048603.1 | - | 1 - 2 | 133 | 101 | |
BC117936.1 | - | 1 - 2 | 87 | 40097789 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |