Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290728.1 | NP_001277657.1 | 1 - 2 | 158 | 100 | |
NM_177704.3 | NP_808372.1 | 1 - 2 | 158 | 100 | ||
GenBank mRNA | AB098160.1 | - | 1 - 2 | 157 | 100 | |
AB098161.1 | - | 1 - 2 | 157 | 100 | ||
BC150158.1 | - | 1 - 2 | 161 | 40141504 | 100 | |
BC150480.1 | - | 1 - 2 | 161 | 40141505 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |