Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164233.1 | NP_001157705.1 | 4 - 5 | 918 | 98 | |
NM_027584.1 | NP_081860.1 | 4 - 5 | 918 | 98 | ||
GenBank mRNA | AK014872.1 | - | 4 - 5 | 919 | 98 | |
AK133206.1 | - | 4 - 5 | 787 | 98 | ||
BC112423.1 | - | 4 - 5 | 888 | 40062055 | 98 | |
BC113131.1 | - | 4 - 5 | 888 | 40062053 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |