Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028140.4 | NP_082416.2 | 1 - 2 | 213 | 84 | |
GenBank mRNA | AK032080.1 | - | 1 - 2 | 234 | 84 | |
AK035291.1 | - | 1 - 2 | 176 | 84 | ||
AK077957.1 | - | 1 - 2 | 214 | 84 | ||
BC113183.1 | - | 1 - 2 | 144 | 40061891 | 84 | |
BC113790.1 | - | 1 - 2 | 144 | 40061889 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |