Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027439.4 | NP_081715.1 | 7 - 8 | 884 | 85 | |
GenBank mRNA | AB192471.2 | - | 7 - 8 | 760 | 85 | |
AK017482.1 | - | 7 - 8 | 816 | 85 | ||
AK029405.1 | - | 7 - 8 | 787 | 85 | ||
AK076495.1 | - | 7 - 8 | 829 | 85 | ||
AK151735.1 | - | 7 - 8 | 816 | 85 | ||
AK168139.1 | - | 7 - 8 | 841 | 85 | ||
AW985491.1 | - | 4 - 5 | 451 | 85 | ||
BC014706.1 | - | 7 - 8 | 755 | 4457079 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |