Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033039.2 | NP_001028211.1 | 2 - 3 | 823 | 77 | |
GenBank mRNA | AK076812.1 | - | 2 - 3 | 546 | 77 | |
AK140290.1 | - | 2 - 3 | 824 | 77 | ||
BC141196.1 | - | 2 - 3 | 736 | 9055808 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |