Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027357.2 | NP_081633.1 | 18 - 19 | 2283 | 68 | |
GenBank mRNA | AK010596.1 | - | 2 - 3 | 229 | 68 | |
AK087947.1 | - | 18 - 19 | 2234 | 68 | ||
AK153386.1 | - | 18 - 19 | 2284 | 68 | ||
AK159123.1 | - | 18 - 19 | 2283 | 68 | ||
AK159750.1 | - | 18 - 19 | 2262 | 68 | ||
BC018172.1 | - | 4 - 5 | 332 | 68 | ||
BC022955.1 | - | 18 - 19 | 2173 | 68 | ||
BC025802.1 | - | 18 - 19 | 2187 | 68 | ||
BC029319.1 | - | 18 - 19 | 2220 | 68 | ||
BC031587.1 | - | 18 - 19 | 2214 | 68 | ||
BC032068.1 | - | 18 - 19 | 2214 | 68 | ||
BC039937.1 | - | 18 - 19 | 2260 | 68 | ||
BC089028.1 | - | 18 - 19 | 2230 | 68 | ||
BC138526.1 | - | 18 - 19 | 2246 | 8861546 | 68 | |
BC138527.1 | - | 18 - 19 | 2246 | 8861547 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |