Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133821.3 | NP_598582.3 | 8 - 9 | 2712 | 79 | |
GenBank mRNA | AB093251.1 | - | 8 - 9 | 1471 | 79 | |
BC059254.1 | - | 8 - 9 | 1372 | 6406203 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |