Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500008589.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008589.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008589.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008586.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008586.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008586.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008587.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008587.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008587.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008590.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008590.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008590.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008593.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008593.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008593.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008591.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008591.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008591.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008592.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008592.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008592.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008594.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008594.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008594.mm.1 |
TC1500001085.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500008595.mm.1 |
NM_145625.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008595.mm.1 |
NM_145625.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500008595.mm.1 |
TC1500001085.mm.1 |
MTA TC |