Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080925.1 | NP_001074394.1 | 7 - 8 | 1576 | 107 | |
GenBank mRNA | BC080279.1 | - | 6 - 7 | 644 | 107 | |
BC130266.1 | - | 8 - 9 | 852 | 40143028 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |