Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159748.1 | NP_001153220.1 | 2 - 3 | 462 | 145 | |
NM_011459.4 | NP_035589.1 | 2 - 3 | 462 | 145 | ||
GenBank mRNA | AK032659.1 | - | 2 - 3 | 462 | 145 | |
AK036811.1 | - | 2 - 3 | 398 | 145 | ||
AK048230.1 | - | 2 - 3 | 422 | 145 | ||
BC011076.1 | - | 2 - 3 | 441 | 145 | ||
BC025121.1 | - | 2 - 3 | 452 | 4487039 | 145 | |
U96703.2 | - | 2 - 3 | 453 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |