Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007689.4 | NP_031715.1 | 2 - 3 | 959 | 83 | |
GenBank mRNA | AK158297.1 | - | 2 - 3 | 987 | 83 | |
AK160489.1 | - | 2 - 3 | 555 | 83 | ||
BC012672.1 | - | 2 - 3 | 998 | 4192855 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |