Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081434.1 | NP_001074903.1 | 20 - 21 | 2328 | 59 | |
GenBank mRNA | AK017095.1 | - | 2 - 3 | 349 | 59 | |
AK154677.1 | - | 20 - 21 | 2366 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |