Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007622.3 | NP_031648.1 | 2 - 3 | 392 | 71 | |
GenBank mRNA | AK014215.1 | - | 2 - 3 | 368 | 71 | |
AK017608.1 | - | 2 - 3 | 281 | 71 | ||
AK157710.1 | - | 2 - 3 | 394 | 71 | ||
BC055290.1 | - | 2 - 3 | 335 | 6313666 | 71 | |
X56690.1 | - | 1 - 2 | 137 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |