Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205353.1 | NP_001192282.1 | 9 - 10 | 1106 | 79 | |
NM_172611.4 | NP_766199.1 | 9 - 10 | 1008 | 79 | ||
GenBank mRNA | AK036832.1 | - | 9 - 10 | 997 | 79 | |
AK129209.1 | - | 9 - 10 | 910 | 79 | ||
AK142266.1 | - | 9 - 10 | 961 | 79 | ||
BC021523.1 | - | 5 - 6 | 382 | 79 | ||
BC085148.1 | - | 9 - 10 | 1064 | 79 | ||
BC139228.1 | - | 9 - 10 | 997 | 8862250 | 79 | |
BC139229.1 | - | 9 - 10 | 997 | 8862251 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |