Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025730.3 | NP_080006.3 | 44 - 45 | 6634 | 112 | |
GenBank mRNA | AK014886.1 | - | 5 - 6 | 693 | 112 | |
AK014938.1 | - | 5 - 6 | 706 | 112 | ||
AY792512.1 | - | 44 - 45 | 6577 | 112 | ||
BC034074.1 | - | 10 - 11 | 1517 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |