Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10606817 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10606818 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10606819 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1431851_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10606816 |
- |
- |
Mouse Genome 430 2.0 Array |
1431851_at |
- |
- |
Mouse Genome 430 2.0 Array |
1431851_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
75829_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
75829_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00018868.mm.1 |
NM_029459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018868.mm.1 |
NM_029459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018868.mm.1 |
TC0X00003013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018870.mm.1 |
NM_029459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018870.mm.1 |
NM_029459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018870.mm.1 |
TC0X00003013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018869.mm.1 |
NM_029459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018869.mm.1 |
NM_029459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018869.mm.1 |
TC0X00003013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018867.mm.1 |
NM_029459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018867.mm.1 |
NM_029459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018867.mm.1 |
TC0X00003013.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1431851_at |
- |
- |
Mouse Expression Set 430 |
1431851_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1431851_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1431851_at |
- |
- |