Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027427.2 | NP_081703.1 | 8 - 9 | 728 | 57 | |
GenBank mRNA | AK051568.1 | - | 8 - 9 | 729 | 57 | |
AK146331.1 | - | 8 - 9 | 691 | 57 | ||
AK166550.1 | - | 8 - 9 | 721 | 57 | ||
BC043716.1 | - | 2 - 3 | 56 | 57 | ||
BC137591.1 | - | 8 - 9 | 702 | 8860610 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |