Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028797.1 | NP_083073.1 | 1 - 2 | 72 | 98 | |
GenBank mRNA | AK005675.1 | - | 1 - 2 | 50 | 98 | |
BU945975.1 | - | 1 - 2 | 75 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |