Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163389.1 | NP_001156861.1 | 3 - 4 | 255 | 114 | |
NM_028965.4 | NP_083241.1 | 4 - 5 | 315 | 114 | ||
GenBank mRNA | AK037747.1 | - | 2 - 3 | 2699 | 114 | |
AK042409.1 | - | 3 - 4 | 231 | 114 | ||
AK076332.1 | - | 4 - 5 | 291 | 114 | ||
BC014719.1 | - | 3 - 4 | 232 | 4236099 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |