Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144838.1 | NP_659087.1 | 9 - 10 | 916 | 77 | |
GenBank mRNA | AK049820.1 | - | 9 - 10 | 954 | 77 | |
AK082080.1 | - | 9 - 10 | 806 | 77 | ||
BC017611.1 | - | 9 - 10 | 916 | 4527683 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |