Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025445.4 | NP_079721.2 | 1 - 2 | 167 | 89 | |
GenBank mRNA | AK007732.1 | - | 1 - 2 | 84 | 89 | |
AK028990.1 | - | 1 - 2 | 168 | 89 | ||
AK075788.1 | - | 1 - 2 | 156 | 89 | ||
AK169107.1 | - | 1 - 2 | 77 | 89 | ||
BC004081.1 | - | 1 - 2 | 103 | 3592719 | 89 | |
BC060369.1 | - | 1 - 2 | 94 | 6310619 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |