Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027026.2 | NP_081302.2 | 4 - 5 | 551 | 71 | |
GenBank mRNA | AK005668.1 | - | 4 - 5 | 527 | 71 | |
AK158517.1 | - | 4 - 5 | 598 | 71 | ||
BC022723.1 | - | 4 - 5 | 551 | 4503387 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |