Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146027.2 | NP_666139.1 | 1 - 2 | 110 | 111 | |
GenBank mRNA | AK075869.1 | - | 1 - 2 | 74 | 111 | |
CN691874.1 | - | 1 - 2 | 110 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |