Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080127.1 | NP_001073596.1 | 6 - 7 | 846 | 83 | |
NM_001080128.1 | NP_001073597.1 | 5 - 6 | 665 | 83 | ||
NM_009070.2 | NP_033096.2 | 7 - 8 | 968 | 83 | ||
GenBank mRNA | AK146443.1 | - | 5 - 6 | 664 | 83 | |
AK151150.1 | - | 6 - 7 | 836 | 83 | ||
AK165905.1 | - | 5 - 6 | 652 | 83 | ||
BC002061.1 | - | 6 - 7 | 834 | 3485311 | 83 | |
BC008089.1 | - | 3 - 4 | 403 | 83 | ||
X70067.1 | - | 6 - 7 | 853 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |