Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177375.1 | NP_796349.1 | 1 - 2 | 373 | 115 | |
GenBank mRNA | AB232621.2 | - | 1 - 2 | 209 | 115 | |
AK043912.1 | - | 1 - 2 | 338 | 115 | ||
AK080607.1 | - | 353 | 115 | |||
BC094931.1 | - | 1 - 2 | 281 | 6436242 | 115 | |
BC118018.1 | - | 1 - 2 | 514 | 40103722 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |