Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028792.3 | NP_083068.1 | 1 - 2 | 1145 | 76 | |
GenBank mRNA | AK004913.1 | - | 1 - 2 | 1094 | 76 | |
AK154799.1 | - | 1 - 2 | 1194 | 76 | ||
AK211745.1 | - | 1 - 2 | 271 | 76 | ||
BC006928.1 | - | 1 - 2 | 293 | 2655826 | 76 | |
BC086769.1 | - | 1 - 2 | 265 | 5707990 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |