Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161712.1 | NP_001155184.1 | 5 - 6 | 781 | 67 | |
NM_013847.4 | NP_038875.3 | 5 - 6 | 781 | 67 | ||
GenBank mRNA | AF093403.1 | - | 5 - 6 | 739 | 67 | |
AK013138.1 | - | 5 - 6 | 736 | 67 | ||
AK075782.1 | - | 5 - 6 | 721 | 67 | ||
AK076469.1 | - | 5 - 6 | 754 | 67 | ||
AK087433.1 | - | 5 - 6 | 740 | 67 | ||
BC024107.1 | - | 5 - 6 | 781 | 5125361 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |