Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080981.1 | NP_001074450.1 | 7 - 8 | 817 | 81 | |
GenBank mRNA | AK014820.1 | - | 5 - 6 | 1575 | 81 | |
BC049928.1 | - | 5 - 6 | 1547 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |