Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Troap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162506.1 | NP_001155978.1 | 6 - 7 | 813 | 71 | |
NM_030159.1 | NP_084435.1 | 6 - 7 | 838 | 71 | ||
GenBank mRNA | AK050381.1 | - | 6 - 7 | 834 | 71 | |
AK077509.1 | - | 6 - 7 | 803 | 71 | ||
BC100405.1 | - | 6 - 7 | 767 | 71 | ||
BC145227.1 | - | 6 - 7 | 1211 | 9053779 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |