Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011950.2 | NP_036080.2 | 7 - 8 | 703 | 96 | |
GenBank mRNA | AK146993.1 | - | 7 - 8 | 691 | 96 | |
BC001992.1 | - | 7 - 8 | 672 | 3492903 | 96 | |
U81823.1 | - | 7 - 8 | 660 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |