Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163691.1 | NP_001157163.1 | 26 - 27 | 5274 | 92 | |
NM_021415.4 | NP_067390.4 | 26 - 27 | 5274 | 92 | ||
GenBank mRNA | AK122452.1 | - | 15 - 16 | 2247 | 92 | |
AK135951.1 | - | 8 - 9 | 1021 | 92 | ||
AK147242.1 | - | 26 - 27 | 5275 | 92 | ||
BC138026.1 | - | 26 - 27 | 5113 | 8861045 | 92 | |
BU612176.1 | - | 4 - 5 | 515 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |