Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046962.1 | - | 1 | 1675 | 64 | |
AK053696.1 | - | 1 | 1544 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |