Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mslnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177822.3 | NP_808490.1 | 8 - 9 | 1103 | 84 | |
GenBank mRNA | AK028891.1 | - | 8 - 9 | 1103 | 84 | |
BC052484.1 | - | 7 - 8 | 839 | 6312790 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |