Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164177.1 | NP_001157649.1 | 3 - 4 | 401 | 71 | |
NM_016883.4 | NP_058579.2 | 3 - 4 | 401 | 71 | ||
GenBank mRNA | AK009068.1 | - | 3 - 4 | 380 | 71 | |
AK018233.1 | - | 3 - 4 | 379 | 71 | ||
AK136400.1 | - | 3 - 4 | 380 | 71 | ||
BC026931.1 | - | 3 - 4 | 368 | 3987112 | 71 | |
BC056196.1 | - | 3 - 4 | 386 | 30019985 | 71 | |
CD564300.1 | - | 3 - 4 | 368 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |