Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700014100.mm.1 |
NM_027644.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014100.mm.1 |
NM_027644.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014100.mm.1 |
TC1700001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014103.mm.1 |
NM_027644.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014103.mm.1 |
NM_027644.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014103.mm.1 |
TC1700001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014106.mm.1 |
NM_027644.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014106.mm.1 |
NM_027644.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014106.mm.1 |
TC1700001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700014102.mm.1 |
NM_027644.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014102.mm.1 |
NM_027644.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700014102.mm.1 |
TC1700001608.mm.1 |
MTA TC |