Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700003024.mm.1 |
NM_001197024.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003024.mm.1 |
NM_001197024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003024.mm.1 |
TC1700000366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003021.mm.1 |
NM_001197024.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003021.mm.1 |
NM_001197024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003021.mm.1 |
TC1700000366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003022.mm.1 |
NM_001197024.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003022.mm.1 |
NM_001197024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003022.mm.1 |
TC1700000366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700003025.mm.1 |
NM_001197024.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003025.mm.1 |
NM_001197024.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700003025.mm.1 |
TC1700000366.mm.1 |
MTA TC |