Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Numb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136075.2 | NP_001129547.1 | 4 - 5 | 322 | 131 | |
NM_001272055.1 | NP_001258984.1 | 3 - 4 | 289 | 131 | ||
NM_001272056.1 | NP_001258985.1 | 5 - 6 | 519 | 131 | ||
NM_010949.2 | NP_035079.1 | 4 - 5 | 322 | 131 | ||
NR_073563.1 | 6 - 7 | 636 | 131 | |||
NR_073564.1 | 7 - 8 | 669 | 131 | |||
GenBank mRNA | AF169191.1 | - | 3 - 4 | 274 | 131 | |
AF169192.1 | - | 4 - 5 | 307 | 131 | ||
AF170709.1 | - | 4 - 5 | 307 | 131 | ||
AK004553.1 | - | 6 - 7 | 544 | 131 | ||
BC050108.1 | - | 7 - 8 | 579 | 131 | ||
U70674.1 | - | 5 - 6 | 440 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |