Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165929.1 | NP_001159401.1 | 12 - 13 | 1250 | 74 | |
GenBank mRNA | AK140238.1 | - | 11 - 12 | 1718 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons |