Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700016605.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016605.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016605.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016605.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016605.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016610.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016610.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016610.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016610.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016610.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016602.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016602.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016602.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016602.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016602.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016604.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016604.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016604.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016604.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016604.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016601.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016601.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016601.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016601.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016601.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016608.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016608.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016608.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016608.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016608.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016599.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016599.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016599.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016599.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016599.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016606.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016606.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016606.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016606.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016606.mm.1 |
TC1700001798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016607.mm.1 |
NM_027060.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016607.mm.1 |
NM_027060.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016607.mm.1 |
NM_172618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016607.mm.1 |
NM_172618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016607.mm.1 |
TC1700001798.mm.1 |
MTA TC |