Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277938.1 | NP_001264867.1 | 2 - 3 | 356 | 65 | |
NM_001277939.1 | NP_001264868.1 | 2 - 3 | 356 | 65 | ||
NM_001277941.1 | NP_001264870.1 | 2 - 3 | 356 | 65 | ||
NM_001277942.1 | NP_001264871.1 | 2 - 3 | 356 | 65 | ||
NM_011235.4 | NP_035365.1 | 2 - 3 | 356 | 65 | ||
NR_102717.1 | 2 - 3 | 356 | 65 | |||
NR_102718.1 | 2 - 3 | 356 | 65 | |||
GenBank mRNA | AB007040.1 | - | 2 - 3 | 321 | 65 | |
AB052828.1 | - | 2 - 3 | 321 | 65 | ||
AB052829.1 | - | 2 - 3 | 321 | 65 | ||
AB052830.1 | - | 2 - 3 | 321 | 65 | ||
AB052833.1 | - | 2 - 3 | 321 | 65 | ||
AB052834.1 | - | 2 - 3 | 321 | 65 | ||
AF034955.1 | - | 2 - 3 | 346 | 65 | ||
AK017835.1 | - | 2 - 3 | 339 | 65 | ||
AK138553.1 | - | 2 - 3 | 348 | 65 | ||
AK147759.1 | - | 2 - 3 | 331 | 65 | ||
BC049136.1 | - | 2 - 3 | 321 | 6832014 | 65 | |
CJ141615.1 | - | 2 - 3 | 356 | 65 | ||
EU627687.1 | - | 2 - 3 | 346 | 65 | ||
EU627688.1 | - | 2 - 3 | 346 | 65 | ||
Y15570.1 | - | 2 - 3 | 342 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |