Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285890.1 | NP_001272819.1 | 12 - 13 | 1703 | 80 | |
NM_008800.2 | NP_032826.1 | 13 - 14 | 1720 | 80 | ||
GenBank mRNA | AK004772.1 | - | 12 - 13 | 1310 | 80 | |
L01695.1 | - | 13 - 14 | 1463 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |