Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080118.1 | NP_001073587.1 | 13 - 14 | 1309 | 102 | |
NM_013634.2 | NP_038662.2 | 13 - 14 | 1174 | 102 | ||
NM_134027.2 | NP_598788.2 | 13 - 14 | 1309 | 102 | ||
GenBank mRNA | AF000294.1 | - | 13 - 14 | 1174 | 102 | |
AF332073.1 | - | 12 - 13 | 1054 | 102 | ||
AF332074.1 | - | 12 - 13 | 1054 | 102 | ||
AK049203.1 | - | 13 - 14 | 1300 | 102 | ||
AK147199.1 | - | 13 - 14 | 1286 | 102 | ||
BC021440.1 | - | 13 - 14 | 1280 | 102 | ||
BC079636.1 | - | 13 - 14 | 1279 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |