Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083114.1 | NP_001076583.1 | 2 - 3 | 205 | 91 | |
NM_001285863.1 | NP_001272792.1 | 2 - 3 | 334 | 91 | ||
NM_175363.4 | NP_780572.1 | 2 - 3 | 205 | 91 | ||
GenBank mRNA | AK081606.1 | - | 2 - 3 | 198 | 91 | |
AK150904.1 | - | 2 - 3 | 156 | 91 | ||
AK158488.1 | - | 2 - 3 | 335 | 91 | ||
AK160138.1 | - | 2 - 3 | 178 | 91 | ||
AK168443.1 | - | 2 - 3 | 206 | 91 | ||
BC150854.2 | - | 1 - 2 | 109 | 9087765 | 91 | |
BY006733.1 | - | 2 - 3 | 206 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |