Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2042319 |
AK004984.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2042319 |
BC061152.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2042319 |
BC087906.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2042319 |
NM_023623.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
762988 |
BC046770.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
762988 |
BC059941.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
762988 |
NM_008990.2 |
refseq_mrna_mouse_NM |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P102886 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2042319 |
AK004984.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2042319 |
BC061152.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2042319 |
BC087906.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2042319 |
NM_023623.2 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P102886 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P102886 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12415327 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12415329 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12415315 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12415323 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12415311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384497 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384480 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384455 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384530 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30384458 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12415334 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12415325 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384521 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384473 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384510 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384500 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384528 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12415323 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384458 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12323265 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12415329 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384480 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12415313 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12394958 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384522 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12415326 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384454 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12394987 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384488 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384526 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384464 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384455 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384465 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384518 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12394940 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384529 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384459 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384469 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384524 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384478 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12323264 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11927091 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384472 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384503 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384483 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384494 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384476 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P12394962 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P30384514 |
- |
- |