Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027905.2 | NP_082181.2 | 3 - 4 | 956 | 68 | |
GenBank mRNA | AK005048.1 | - | 3 - 4 | 956 | 68 | |
AK054432.1 | - | 3 - 4 | 918 | 68 | ||
AK161059.1 | - | 3 - 4 | 907 | 68 | ||
BC117817.1 | - | 4 - 5 | 471 | 68 | ||
BC117818.1 | - | 4 - 5 | 504 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |